siRNA Details
| Gene Symbol: Slc17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133554.2 | NP_598238.2 | 851 | |||
| GenBank mRNA | AY102171.1 | - | 719 | |||
| BC078748.1 | - | 851 | 7109368 | |||
| U28504.1 | - | 750 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a1| Gene Symbol: Slc17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133554.2 | NP_598238.2 | 851 | |||
| GenBank mRNA | AY102171.1 | - | 719 | |||
| BC078748.1 | - | 851 | 7109368 | |||
| U28504.1 | - | 750 | ||||