siRNA Details
| Gene Symbol: Mpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133561.1 | NP_598245.1 | 5 | 497 | ||
| GenBank mRNA | AF304429.1 | - | 5 | 497 | ||
| BC097287.1 | - | 5 | 513 | 7445460 | ||
| FQ219877.1 | - | 5 | 551 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpc1| Gene Symbol: Mpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133561.1 | NP_598245.1 | 5 | 497 | ||
| GenBank mRNA | AF304429.1 | - | 5 | 497 | ||
| BC097287.1 | - | 5 | 513 | 7445460 | ||
| FQ219877.1 | - | 5 | 551 | |||