siRNA Details
| Gene Symbol: Angptl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133569.1 | NP_598253.1 | 4 | 1472 | ||
| GenBank mRNA | AF159049.1 | - | 4 | 1472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angptl2| Gene Symbol: Angptl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133569.1 | NP_598253.1 | 4 | 1472 | ||
| GenBank mRNA | AF159049.1 | - | 4 | 1472 | ||