siRNA Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133571.1 | NP_598255.1 | 2 | 450 | ||
| XM_006243702.2 | XP_006243764.1 | 2 | 675 | |||
| XM_006243703.1 | XP_006243765.1 | 3 | 223 | |||
| GenBank mRNA | AY724516.1 | - | 2 | 570 | ||
| D16236.1 | - | 2 | 450 | |||

Cdc25a
Genomic Map