siRNA Details
Gene Symbol: Il1rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133575.1 | NP_598259.1 | 3 | 186 | ||
GenBank mRNA | U49066.1 | - | 3 | 186 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il1rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133575.1 | NP_598259.1 | 3 | 186 | ||
GenBank mRNA | U49066.1 | - | 3 | 186 |