siRNA Details
| Gene Symbol: Il1rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133575.1 | NP_598259.1 | 4 | 434 | ||
| GenBank mRNA | U49066.1 | - | 4 | 434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1rl2| Gene Symbol: Il1rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133575.1 | NP_598259.1 | 4 | 434 | ||
| GenBank mRNA | U49066.1 | - | 4 | 434 | ||