siRNA Details
Gene Symbol: Chrnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133597.1 | NP_598281.1 | 4 | 515 | ||
GenBank mRNA | AY574259.1 | - | 4 | 338 | ||
J04636.1 | - | 4 | 515 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chrnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133597.1 | NP_598281.1 | 4 | 515 | ||
GenBank mRNA | AY574259.1 | - | 4 | 338 | ||
J04636.1 | - | 4 | 515 |