siRNA Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133624.2 | NP_598308.1 | 2 | 245 | ||
| GenBank mRNA | BC086991.1 | - | 2 | 207 | 7191761 | |
| M80367.1 | - | 2 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbp2| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133624.2 | NP_598308.1 | 2 | 245 | ||
| GenBank mRNA | BC086991.1 | - | 2 | 207 | 7191761 | |
| M80367.1 | - | 2 | 246 | |||