siRNA Details
| Gene Symbol: Clk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134340.1 | NP_599167.1 | 2, 3 | 216 | ||
| XM_006243146.2 | XP_006243208.1 | 2, 3 | 1161 | |||
| XM_006243149.2 | XP_006243211.1 | 2, 3 | 178 | |||
| GenBank mRNA | BC070891.1 | - | 2, 3 | 192 | 7099115 | |
| X94351.1 | - | 2, 3 | 216 | |||

Clk3
Genomic Map