siRNA Details
| Gene Symbol: Sh2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048180.1 | NP_001041645.1 | 1 | 276 | ||
| NM_134456.3 | NP_604451.2 | 2 | 576 | |||
| XM_006230351.2 | XP_006230413.1 | 1 | 2137 | |||
| XM_006230352.2 | XP_006230414.1 | 1 | 2137 | |||
| XM_006230353.2 | XP_006230415.1 | 2 | 551 | |||
| XM_006230354.2 | XP_006230416.1 | 2 | 1457 | |||
| XM_006230355.2 | XP_006230417.1 | 1 | 2136 | |||
| GenBank mRNA | AF047577.1 | - | 1 | 276 | ||
| U57391.1 | - | 2 | 619 | |||

Sh2b1
Genomic Map