siRNA Details
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134459.1 | NP_604454.1 | 2, 3 | 200 | ||
XM_006251676.2 | XP_006251738.1 | 2, 3 | 236 | |||
XM_008770596.1 | XP_008768818.1 | 2, 3 | 226 | |||
GenBank mRNA | AB031014.1 | - | 2, 3 | 200 | ||
AF481858.1 | - | 2, 3 | 147 |
