siRNA Details
| Gene Symbol: Cd99l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134459.1 | NP_604454.1 | 3 | 205 | ||
| XM_006251676.2 | XP_006251738.1 | 3 | 241 | |||
| XM_008770596.1 | XP_008768818.1 | 3 | 231 | |||
| GenBank mRNA | AB031014.1 | - | 3 | 205 | ||
| AF481858.1 | - | 3 | 152 | |||

Cd99l2
Genomic Map