siRNA Details
| Gene Symbol: Dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138519.2 | NP_612528.2 | 8 | 2665 | ||
| XM_006230009.2 | XP_006230071.1 | 8 | 2673 | |||
| GenBank mRNA | AF245040.1 | - | 7 | 2529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dkk3| Gene Symbol: Dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138519.2 | NP_612528.2 | 8 | 2665 | ||
| XM_006230009.2 | XP_006230071.1 | 8 | 2673 | |||
| GenBank mRNA | AF245040.1 | - | 7 | 2529 | ||