siRNA Details
| Gene Symbol: Epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138541.1 | NP_612550.1 | 1, 2 | 142 | ||
| GenBank mRNA | AJ001044.1 | - | 1, 2 | 197 | ||
| BC061787.1 | - | 1, 2 | 146 | |||
| BC072691.1 | - | 1, 2 | 166 | 7098920 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epcam| Gene Symbol: Epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138541.1 | NP_612550.1 | 1, 2 | 142 | ||
| GenBank mRNA | AJ001044.1 | - | 1, 2 | 197 | ||
| BC061787.1 | - | 1, 2 | 146 | |||
| BC072691.1 | - | 1, 2 | 166 | 7098920 | ||