siRNA Details
Gene Symbol: Syt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138835.1 | NP_620190.1 | 3, 4 | 456 | ||
XM_008760071.1 | XP_008758293.1 | 3, 4 | 384 | |||
XM_008760072.1 | XP_008758294.1 | 4, 5 | 430 | |||
GenBank mRNA | U71294.1 | - | 3, 4 | 456 |
