siRNA Details
| Gene Symbol: Gpm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138846.1 | NP_620201.1 | 2, 3 | 492 | ||
| XM_006256849.2 | XP_006256911.1 | 3, 4 | 1031 | |||
| XM_006256850.2 | XP_006256912.1 | 3, 4 | 1033 | |||
| XM_006256851.2 | XP_006256913.1 | 2, 3 | 555 | |||
| XM_008773151.1 | XP_008771373.1 | 2, 3 | 544 | |||
| GenBank mRNA | AB036421.1 | - | 2, 3 | 492 | ||

Gpm6b
Genomic Map