siRNA Details
| Gene Symbol: Gpm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138846.1 | NP_620201.1 | 7 | 1058 | ||
| GenBank mRNA | AB036421.1 | - | 7 | 1058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpm6b| Gene Symbol: Gpm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138846.1 | NP_620201.1 | 7 | 1058 | ||
| GenBank mRNA | AB036421.1 | - | 7 | 1058 | ||