siRNA Details
| Gene Symbol: Syt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138849.1 | NP_620204.1 | 4 | 293 | ||
| XM_006230101.2 | XP_006230163.1 | 4 | 388 | |||
| XM_006230102.2 | XP_006230164.1 | 4 | 273 | |||
| XM_006230104.1 | XP_006230166.1 | 4 | 487 | |||
| XM_008759675.1 | XP_008757897.1 | 2 | 753 | |||
| GenBank mRNA | U30831.1 | - | 4 | 293 | ||

Syt17
Genomic Map