siRNA Details
| Gene Symbol: Nbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138873.2 | NP_620228.1 | 4 | 376 | ||
| XM_006237916.1 | XP_006237978.1 | 4 | 419 | |||
| GenBank mRNA | AF218575.1 | - | 4 | 380 | ||
| BC085700.1 | - | 4 | 344 | 7130978 | ||

Nbn
Genomic Map