siRNA Details
| Gene Symbol: C1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138900.1 | NP_620255.1 | 3 | 425 | ||
| GenBank mRNA | BC062042.1 | - | 3 | 392 | 5598051 | |
| D88250.1 | - | 3 | 425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1s| Gene Symbol: C1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138900.1 | NP_620255.1 | 3 | 425 | ||
| GenBank mRNA | BC062042.1 | - | 3 | 392 | 5598051 | |
| D88250.1 | - | 3 | 425 | |||