siRNA Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138911.3 | NP_620266.1 | 9 | 1159 | ||
| GenBank mRNA | BC061529.1 | - | 9 | 1160 | 6887497 | |
| FQ220758.1 | - | 6 | 681 | |||
| Y15068.1 | - | 8 | 1094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stip1| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138911.3 | NP_620266.1 | 9 | 1159 | ||
| GenBank mRNA | BC061529.1 | - | 9 | 1160 | 6887497 | |
| FQ220758.1 | - | 6 | 681 | |||
| Y15068.1 | - | 8 | 1094 | |||