siRNA Details
| Gene Symbol: Fbxo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138917.3 | NP_620272.3 | 5 | 821 | ||
| XM_006239372.2 | XP_006239434.1 | 6 | 1125 | |||
| XM_006239373.2 | XP_006239435.1 | 6 | 1058 | |||
| XM_006239374.2 | XP_006239436.1 | 6 | 985 | |||
| XM_006239375.2 | XP_006239437.1 | 6 | 989 | |||
| GenBank mRNA | AF393484.1 | - | 5 | 821 | ||
| BC063148.1 | - | 5 | 823 | 6921385 | ||
| FQ232698.1 | - | 6 | 891 | |||

Fbxo6
Genomic Map