siRNA Details
| Gene Symbol: Rnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270762.1 | NP_001257691.1 | 3 | 296 | ||
| NM_001270763.1 | NP_001257692.1 | 3 | 302 | |||
| NM_139105.3 | NP_620805.2 | 3 | 311 | |||
| XM_008759970.1 | XP_008758192.1 | 3 | 860 | |||
| GenBank mRNA | AB296102.1 | - | 2 | 178 | ||
| BC070501.1 | - | 3 | 218 | 7096144 | ||
| FQ233708.1 | - | 3 | 236 | |||
| FQ234889.1 | - | 3 | 235 | |||
| X62528.1 | - | 3 | 296 | |||

Rnh1
Genomic Map