siRNA Details
| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139190.1 | NP_631929.1 | 2695 | |||
| GenBank mRNA | AY078385.1 | - | 2695 | |||
| BC081722.1 | - | 2663 | 7124320 | |||
| Gene Symbol: LOC100912282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750418.3 | XP_003750466.1 | 15 | 2757 | ||

Calcoco1
Genomic Map