siRNA Details
| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139194.2 | NP_631933.2 | 2 | 202 | ||
| GenBank mRNA | AF104034.1 | - | 2 | 134 | ||
| CB810875.1 | - | 2 | 150 | |||
| D26113.1 | - | 2 | 202 | |||
| FQ233018.1 | - | 2 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fas| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139194.2 | NP_631933.2 | 2 | 202 | ||
| GenBank mRNA | AF104034.1 | - | 2 | 134 | ||
| CB810875.1 | - | 2 | 150 | |||
| D26113.1 | - | 2 | 202 | |||
| FQ233018.1 | - | 2 | 190 | |||