siRNA Details
| Gene Symbol: TOPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007027.3 | NP_008958.2 | 6 | 784 | ||
| XM_005247076.3 | XP_005247133.1 | 6 | 1271 | |||
| XM_011512357.2 | XP_011510659.1 | 6 | 855 | |||
| XM_017005636.1 | XP_016861125.1 | 6 | 854 | |||
| XM_017005637.1 | XP_016861126.1 | 3 | 390 | |||
| GenBank mRNA | AB019397.1 | - | 6 | 731 | ||
| AK302584.1 | - | 6 | 750 | |||
| BC126209.1 | - | 5 | 674 | 8991925 | ||
| BC151237.1 | - | 6 | 737 | 8860504 | ||
| D87448.1 | - | 6 | 737 | |||

TOPBP1
Genomic Map