siRNA Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145098.2 | NP_659566.1 | 5 | 1086 | ||
| XM_006255825.2 | XP_006255887.1 | 5 | 1279 | |||
| XM_006255826.2 | XP_006255888.1 | 4 | 1064 | |||
| XM_006255827.2 | XP_006255889.1 | 4 | 1064 | |||
| XM_008772598.1 | XP_008770820.1 | 4 | 1064 | |||
| GenBank mRNA | AF016296.1 | - | 4 | 690 | ||
| AF018957.1 | - | 4 | 543 | |||
| BC085689.1 | - | 5 | 1086 | 7109697 | ||

Nrp1
Genomic Map