siRNA Details
| Gene Symbol: Mxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145773.1 | NP_665716.1 | 5 | 534 | ||
| GenBank mRNA | FQ234290.1 | - | 5 | 524 | ||
| U45986.1 | - | 5 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mxd3| Gene Symbol: Mxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145773.1 | NP_665716.1 | 5 | 534 | ||
| GenBank mRNA | FQ234290.1 | - | 5 | 524 | ||
| U45986.1 | - | 5 | 534 | |||