siRNA Details
Gene Symbol: Pzp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145779.2 | NP_665722.2 | 1, 2 | 148 | ||
XM_008763310.1 | XP_008761532.1 | 1, 2 | 159 | |||
GenBank mRNA | M77183.1 | - | 1, 2 | 144 | ||
M84000.1 | - | 1, 2 | 187 |
