siRNA Details
| Gene Symbol: Aptx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148889.2 | NP_683687.1 | 6, 7 | 847 | ||
| XM_006238026.2 | XP_006238088.1 | 6, 7 | 1203 | |||
| XM_006238027.2 | XP_006238089.1 | 5, 6 | 1239 | |||
| XM_006238028.2 | XP_006238090.1 | 5, 6 | 1086 | |||
| XM_006238029.2 | XP_006238091.1 | 4, 5 | 420 | |||
| XM_008763607.1 | XP_008761829.1 | 4, 5 | 1098 | |||
| GenBank mRNA | AF398235.1 | - | 5, 6 | 593 | ||
| BC078716.1 | - | 6, 7 | 847 | 7132044 | ||
| FQ214915.1 | - | 2, 3 | 220 | |||

Aptx
Genomic Map