siRNA Details
| Gene Symbol: Nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148891.2 | NP_683689.1 | 8 | 1302 | ||
| GenBank mRNA | AJ492222.1 | - | 8 | 923 | ||
| BC097277.1 | - | 8 | 1302 | 7457194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmt1| Gene Symbol: Nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148891.2 | NP_683689.1 | 8 | 1302 | ||
| GenBank mRNA | AJ492222.1 | - | 8 | 923 | ||
| BC097277.1 | - | 8 | 1302 | 7457194 | ||