siRNA Details
| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152846.1 | NP_690059.1 | 2 | 152 | ||
| GenBank mRNA | AF532184.1 | - | 2 | 152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F7| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152846.1 | NP_690059.1 | 2 | 152 | ||
| GenBank mRNA | AF532184.1 | - | 2 | 152 | ||