siRNA Details
Gene Symbol: Gzma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153468.1 | NP_703198.1 | 3 | 272 | ||
GenBank mRNA | AB082125.1 | - | 3 | 272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gzma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153468.1 | NP_703198.1 | 3 | 272 | ||
GenBank mRNA | AB082125.1 | - | 3 | 272 |