siRNA Details
| Gene Symbol: Myo7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153473.1 | NP_703203.1 | 4 | 517 | ||
| XM_006229742.2 | XP_006229804.1 | 3 | 1323 | |||
| XM_006229743.2 | XP_006229805.1 | 3 | 1322 | |||
| XM_006229744.2 | XP_006229806.1 | 4 | 551 | |||
| GenBank mRNA | AB091825.1 | - | 4 | 517 | ||

Myo7a
Genomic Map