siRNA Details
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153621.1 | NP_705885.1 | 2, 3 | 209 | ||
| XM_006238494.2 | XP_006238556.1 | 3, 4 | 686 | |||
| XM_008763854.1 | XP_008762076.1 | 3, 4 | 770 | |||
| GenBank mRNA | AB072426.1 | - | 2, 3 | 209 | ||

Dab1
Genomic Map