siRNA Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172022.2 | NP_742019.2 | 4 | 1931 | ||
| XM_006235009.2 | XP_006235071.1 | 6 | 3160 | |||
| XM_006235010.2 | XP_006235072.1 | 6 | 3912 | |||
| XM_006235011.2 | XP_006235073.1 | 5 | 3112 | |||
| XM_006235014.2 | XP_006235076.1 | 5 | 1982 | |||
| XM_008762147.1 | XP_008760369.1 | 6 | 2949 | |||
| XM_008762148.1 | XP_008760370.1 | 5 | 2900 | |||
| XM_008762149.1 | XP_008760371.1 | 6 | 2887 | |||
| XM_008762150.1 | XP_008760372.1 | 5 | 2838 | |||
| XM_008762151.1 | XP_008760373.1 | 4 | 1973 | |||
| GenBank mRNA | AJ278801.2 | - | 4 | 1931 | ||

Lzts3
Genomic Map