siRNA Details
| Gene Symbol: Hyal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172040.2 | NP_742037.2 | 2 | 590 | ||
| XM_006243855.2 | XP_006243917.1 | 2 | 611 | |||
| GenBank mRNA | AF034218.1 | - | 2 | 560 | ||
| AF535141.1 | - | 1 | 498 | |||
| BC062410.1 | - | 2 | 590 | 5623650 | ||

Hyal2
Genomic Map