siRNA Details
| Gene Symbol: Fxyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172317.2 | NP_758528.1 | 3, 4 | 103 | ||
| XM_006228818.2 | XP_006228880.1 | 3, 4 | 353 | |||
| XM_006228819.2 | XP_006228881.1 | 3, 4 | 207 | |||
| XM_006228820.2 | XP_006228882.1 | 3, 4 | 343 | |||
| GenBank mRNA | AW142066.1 | - | 2, 3 | 108 | ||
| DY471861.1 | - | 3, 4 | 475 | |||

Fxyd3
Genomic Map