siRNA Details
Gene Symbol: Zfp161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172325.2 | NP_758828.2 | 407 | |||
GenBank mRNA | AY157624.1 | - | 1 | 326 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172325.2 | NP_758828.2 | 407 | |||
GenBank mRNA | AY157624.1 | - | 1 | 326 |