siRNA Details
| Gene Symbol: Aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172327.2 | NP_758830.2 | 3 | 367 | ||
| XM_006230699.2 | XP_006230761.1 | 4 | 565 | |||
| XM_006230700.2 | XP_006230762.1 | 4 | 534 | |||
| GenBank mRNA | AF543560.1 | - | 3 | 401 | ||
| BC089110.1 | - | 3 | 367 | 7311852 | ||
| FQ214270.1 | - | 3 | 475 | |||
| FQ228146.1 | - | 3 | 395 | |||

Aip
Genomic Map