siRNA Details
| Gene Symbol: Wrnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172332.2 | NP_758835.1 | 1, 2 | 1002 | ||
| GenBank mRNA | AY136827.1 | - | 1, 2 | 906 | ||
| BC098652.1 | - | 1, 2 | 1002 | 7306780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wrnip1| Gene Symbol: Wrnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172332.2 | NP_758835.1 | 1, 2 | 1002 | ||
| GenBank mRNA | AY136827.1 | - | 1, 2 | 906 | ||
| BC098652.1 | - | 1, 2 | 1002 | 7306780 | ||