siRNA Details
| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172335.2 | NP_758838.2 | 3 | 381 | ||
| GenBank mRNA | AB051391.1 | - | 3 | 340 | ||
| BC072474.1 | - | 3 | 381 | 7100875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2a| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172335.2 | NP_758838.2 | 3 | 381 | ||
| GenBank mRNA | AB051391.1 | - | 3 | 340 | ||
| BC072474.1 | - | 3 | 381 | 7100875 | ||