siRNA Details
Gene Symbol: Gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172335.2 | NP_758838.2 | 4 | 905 | ||
GenBank mRNA | BC072474.1 | - | 4 | 905 | 7100875 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172335.2 | NP_758838.2 | 4 | 905 | ||
GenBank mRNA | BC072474.1 | - | 4 | 905 | 7100875 |