siRNA Details
| Gene Symbol: Atf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172336.3 | NP_758839.3 | 2 | 832 | ||
| XM_006228980.1 | XP_006229042.1 | 3 | 1110 | |||
| GenBank mRNA | AY123225.1 | - | 2 | 832 | ||
| BC061786.1 | - | 3 | 1110 | 5623784 | ||

Atf5
Genomic Map