siRNA Details
| Gene Symbol: Atf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228980.1 | XP_006229042.1 | 3 | 1502 | ||
| GenBank mRNA | BC061786.1 | - | 3 | 1502 | 5623784 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf5| Gene Symbol: Atf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228980.1 | XP_006229042.1 | 3 | 1502 | ||
| GenBank mRNA | BC061786.1 | - | 3 | 1502 | 5623784 | |