siRNA Details
Gene Symbol: Csn1s2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173106.1 | NP_775129.1 | 227 | |||
GenBank mRNA | AY154894.1 | - | 227 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csn1s2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173106.1 | NP_775129.1 | 227 | |||
GenBank mRNA | AY154894.1 | - | 227 |