siRNA Details
| Gene Symbol: Asic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173135.1 | NP_775158.1 | 10 | 1478 | ||
| GenBank mRNA | AF013598.1 | - | 10 | 1478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asic3| Gene Symbol: Asic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173135.1 | NP_775158.1 | 10 | 1478 | ||
| GenBank mRNA | AF013598.1 | - | 10 | 1478 | ||