siRNA Details
Gene Symbol: Olr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173293.1 | NP_775415.1 | 2 | 1690 | ||
GenBank mRNA | AF079864.1 | - | 2 | 1690 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173293.1 | NP_775415.1 | 2 | 1690 | ||
GenBank mRNA | AF079864.1 | - | 2 | 1690 |