siRNA Details
Gene Symbol: Naglt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176080.2 | NP_788269.1 | 4 | 1730 | ||
GenBank mRNA | AB089802.1 | - | 4 | 1730 | ||
BC098651.1 | - | 4 | 1717 | 7130354 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Naglt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176080.2 | NP_788269.1 | 4 | 1730 | ||
GenBank mRNA | AB089802.1 | - | 4 | 1730 | ||
BC098651.1 | - | 4 | 1717 | 7130354 |