siRNA Details
| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176857.3 | NP_789827.3 | 1 | 563 | ||
| GenBank mRNA | AB045378.2 | - | 1 | 563 | ||
| AF289091.1 | - | 621 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173443, BC006059, BC059209, AK122563, AK030104, and others.

Vcpip1
Genomic Map